Accessories
Clamps for 35/20 spherical joints
Catalog # 60004-978
Supplier: Kemtech America
Supplier #: CC3525
Jump to:
Product Details & Documents
Kemtech pinch clamp designed for spherical joints.
- Stainless steel construction
- Adjustable metal tensioner
For use with 35/20, 35/25 ball joint, or #25 O-ring joints.
Specifications
- Description:Clamps for 35/20 spherical joints