Order Entry
Canada
ContactUsLinkComponent
PurSwab® Round Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
PurSwab® Round Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
  89133-776
Product available on GSA Advantage®
undefined
PurSwab® Round Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
  89133-776
 :  1805-PFRND
Product available on GSA Advantage®

 

  • Environmentally Preferable:
  • Cat. no.:
    89133-776
  • Handle Length:
    126.9 mm (4.996")
  • Head Width:
    23 mm (0.906")
  • Head Length:
    28.6 mm (1.126")
  • Handle Width:
    4.95 mm (0.195")
  • Supplier No.:
    1805-PF RND

 

 

For general cleaning

Made in the USA.

Round foam tip. Polypropylene shaft. Nonsterile.

 : CE marked.
 : Packaged 50 per pack, 20 packs per case.