Specifications
- Garment application:Laboratory
- Length:191/2"
- Color:White
- Height:181/2"
- Cat. no.:CA10010-124
- Supplier No.:TY657SWH0000
Specifications
About this item
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
- Elastic around face opening
- Shoulder length
- Serged seams
- One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Pullover hood design features elastic face opening for good fit around PAPR. Hood extends over shoulders to provide full head protection.