Specifications
- Garment application:Industrial,Paint and lacquer
- Color:White
- Cat. no.:CA80080-916
- Supplier No.:TY273BWH0000
Specifications
About this item
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
- Neck loop
- Waist ties
- Bound seams
- One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
These bib aprons provide enhanced frontal protection. Bound seams and ties at waist allow for one size to fit most.