ContactUsLinkComponent
DEC-QUAT® 100, Veltek Associates
  75795-368
undefined
DEC-QUAT® 100, Veltek Associates
  75795-368
 :  DQ100-07-16Z-01
  • Description:
    DEC-QUAT® 100
  • Size:
    473 ml (16 oz.)
  • Cat. no.:
    75795-368
  • Packaging:
    Plastic bottle, attached trigger spray
  • Sterility:
    Non sterile
  • Form:
    Concentrate, simple mix
  • Supplier No.:
    DQ100-07-16Z01
DEC-QUAT® 100 is a quaternary ammonium solution for use on hard non-porous environmental surfaces.
  • Effective as a broad spectrum hospital disinfectant, fungicide, deodorizer, virucide
  • Food and non-food contact sanitizer effective against a broad spectrum of pathogenic bacteria
  • Available as both sterile and non-sterile
  • Completely traceable and has been validated for sterility and shelf life
This product is designed for use in hospital, medical and health care institutions as a broad-spectrum disinfectant solution on hard, non-porous environmental surfaces such as walls, ceilings, floors, and countertops. Effective against Pseudomonas aeruginosa, Staphylococcus aureus, and Salmonella choleraesuis. Effective against Hepatitis B Virus when contact time is 10 minutes at 1 oz:1 gal of water. Effective against Human Immunodeficiency Virus Type 1 (HIV) after 30 seconds of contact time.
 : Filled sterile by filtration at 0.2 microns. Gamma-irradiated, double-bagged and delivered with lot-specific sterile documentation.