You searched for: Microplate Seals
VWR offers microplate sealing films and foils for the reduction of evaporation in cell growth, PCR, and ELISA. Choose films of polyester for general incubation, polypropylene for heat resistance, or black vinyl for light sensitive samples. Select transparent models for high optical clarity or porous films for gas exchange. Choose ministrips, rolls, or sheet-based films with heat or pressure based seals. Microplate foils for PCR and cold storage feature medical grade adhesive and are ideal when piercing with pipette tips is required.
VWR® Cap Mat
Supplier: VWR International
Re-usable cap mats are available for friction sealing of plates.
Expand 2 Items
VWR® Pierceable, Silicone Microplate Sealing Mats
Supplier: VWR International
Suitable for sealing microplates in incubation and storage applications, silicone sealing mats provide sample protection while minimizing evaporation in a wide range of temperatures.
Expand 4 Items
VWR® Clear-Zone Pierceable Films for Robotics
Supplier: VWR International
With a polyethylene top layer and inert white polypropylene and acrylic adhesive sublayer, this easy-piercing sealing film allows direct sample recovery with single or multichannel pipettors and robotic probes.
Expand 1 Items
VWR® Adhesive Sealing Films for PCR and Robotics
Supplier: VWR International
Features an extra strong adhesive to minimize evaporation, yet removes easily without leaving a residue on the plate.
Expand 2 Items
VWR® Aluminium Foil Seals for PCR
Supplier: VWR International
Precut aluminum foil seals on a roll for PCR applications. Supplied on a roll with holder for ease of use.
Expand 1 Items
VWR® General Purpose Sealing Films
Supplier: VWR International
These general purpose seals are ideal for use with polypropylene plates. They minimize evaporation and adhesive plate residue.
Expand 2 Items
VWR® Sealing Mats for Square Well Plates
Supplier: VWR International
Sealing mats made of EVA (Ethylene Vinyl Acetate), suitable for square well plates.
Expand 2 Items
VWR® Aluminium Foil Seals for Long Term Storage, Chemical Resistant (DMSO)
Supplier: VWR International
Aluminum foils for use with 96-well or 384-well plates. Foils offer excellent chemical resistance (DMSO).
Expand 2 Items
VWR® Silicone Sealing Mats for VWR® Deep Well Plates
Supplier: VWR International
Provide a safe, secure seal to minimize sample loss with VWR® Deep Well Storage Plates.
Expand 4 Items
VWR® Septa Sealing Mat
Supplier: VWR International
This 96-well septa sealing mat is a cost-effective alternative and is compatible with the ABI 3100.
Expand 1 Items
VWR® Adhesive PCR Foil Seals
Supplier: VWR International
These soft, non permeable 63.5 µm thick aluminium foils, with strong medical grade adhesive, eliminate the need for heat sealing devices or mats during thermal cycling.
Expand 2 Items
VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
Supplier: VWR International
Easier to apply and remove as compared to standard molded PCR strip caps.
Expand 1 Items
VWR® Adhesive Sealing Strips for 8-Strip PCR Tubes
Supplier: VWR International
Easier to apply and remove as compared to standard molded PCR strip caps.
Expand 1 Items
VWR® Pierceable Films for ELISA and General Incubation
Supplier: VWR International
These easily-pierceable 94.1 µm polyethylene films with acrylic adhesive allow direct sample recovery with single or multichannel pipettors and robotic probes.
Expand 2 Items
VWR® Sealing Films, Non-Tacky, for qPCR, Storage and Crystallisation
Supplier: VWR International
50 µm polyolefin films with 50 µm inert encapsulated silicone adhesive. Especially suited for qPCR, storage and protein crystallisation applications. The encapsulated silicone adhesive is non tacky until pressed against the sealing surface, at which time adhesive is released only in sealing areas to form the strongest available heat resistant seal around each well on the plate.
Expand 2 Items
VWR® Films with Colour Coded Marking Areas, Polyester
Supplier: VWR International
Identical to polyester films above, but including a colour dot and white writing area on each end tab for plate identification.
Expand 5 Items
VWR® Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Supplier: VWR International
Aluminium foils, 75.7 µm thick, for use with 96- or 384-well plates. Fit inside the rim of raised rim plates. These foils have one partial-width end tab. Available non sterile only.
Expand 2 Items
VWR® Sealing Film, Polyester
Supplier: VWR International
Polyester film, acrylic adhesive. These 49.5 µm thick polyester films minimise evaporation and prevent spillage and contamination between wells.
Expand 2 Items
VWR® Aluminium Foil Seals for Cold Storage
Supplier: VWR International
Aluminium foils with biologically inert adhesive. These foils are easily pierced by manual or automated pipette tips, for sample recovery without generation of aerosols or cross-contamination.
Expand 2 Items
VWR® Films for ELISA and General Incubation, Polyester
Supplier: VWR International
These 77.5 µm thick polyester films provide secure sealing around every well, not just a cover, thus minimising evaporation, preventing spillage and well-to-well contamination, and eliminating 'edge effects' in sensitive ELISA assays.
Expand 3 Items
VWR® Film Seals for Fluorescence and Photoprotection, Black
Supplier: VWR International
These films are 240 µm thick black vinyl with adhesive for use with 96-well, 384-well, and 1536-well black microplates. They fit within the edges of raised-rim plates and may be applied to either the plate top or bottom.
Expand 1 Items
VWR® Precut Pierceable Vinyl Films for Robotics
Supplier: VWR International
These 106.7 µm thick vinyl films with 20 µm adhesive layer are designed for temporary protection of samples in 96-well plates from contamination and evaporation.
Expand 2 Items
VWR® Adhesive PCR Film Seals
Supplier: VWR International
These heat resistant 74.9 µm thick films are designed for thermal cycling applications. Polypropylene films are not pierceable. For PCR applications, where piercing with pipette tips or robotic probes is required for product recovery, use aluminium foil films. For Real-Time PCR applications, where maximum optical clarity is required, use optically-clear polyester films.
Expand 4 Items
VWR® 96-Well EVA and Silicone Microplate Covers
Supplier: VWR International
VWR® 96-well covers are designed for chromatography and high throughput screening (HTS) applications.
Expand 8 Items
VWR® Sealing Film for Automation
Supplier: VWR International
Adhesive sealing film or foil rolls constructed on plastic cores, for use with high throughput automated microplate sealers.
Expand 3 Items
VWR® Silicone Sealing Mat
Supplier: VWR International
Silicone. Seals 96-well round well plates to prevent evaporation during oil-free thermal cycling. Free of RNase, DNase, human DNA. Fits inside lid of thermal cycler. Use from -40[degree]C to 80[degree]C. Does not require application of sealing film. Color: natural.
Expand 1 Items
VWR® Rayon Films for Biological Cultures
Supplier: VWR International
A 139.7 µm thick hydrophobic porous film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth. These rayon films minimise cross-contamination, spillage and evaporation. They allow uniform air and CO₂ exchange for all wells, unlike plate lids which favour exchange for wells near plate edges. Sterile product is packaged in tamper evident bags of 25.
Expand 2 Items
VWR® PCR Films, PE
Supplier: VWR International
These medium-density polyethylene films, 76µm thick with a 46µm thick adhesive layer, are designed for general-purpose microplate sealing and PCR.
Expand 1 Items
VWR® Adhesive Sealing Films for PCR and Storage
Supplier: VWR International
Polyester seals feature a strong adhesive to minimize evaporation during PCR and storage applications.
Expand 1 Items
VWR® Plate Strip Films 2×8, Polyester
Supplier: VWR International
Polyester 2×8 strip films are identical to polyester films in material and adhesive, but designed for sealing only one or two 8-well rows at a time on either strip-well plates or standard plates whenever rows must be selectively protected or accessed.