You searched for: Centrifugal Filters
Centrifugal filters are used in laboratory settings to manipulate peptides, proteins, oligonucleotides, DNA, and RNA samples. The devices successfully clean up labeling and PCR reactions, isolate specific sections, buffer exchange, clarify samples, and remove particulates. Filters with nylon membrane are ideal for prefiltration clean-up of large agarose or acrylamide gel debris prior to purification with an MWCO device. The centrifugal filters will meet concentration, purification, separation and desalting procedure requirements.
VWR® Centrifugal Filters
Supplier: VWR International
Use to: Concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA; clean up labeling and PCR reactions; isolate DNA from agarose gel slices; buffer exchange; separate proteins, oligonucleotides, and RNA from acrylamide gels; clarify samples; or remove particulates.
Expand 11 Items
Jumbosep Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Convenient and reliable concentration, purification, and diafiltration of 15 to 60 ml biological samples.
Expand 11 Items
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Precise, quick recovery of microliter volumes of concentrate from starting volumes up to 5.0 ml.
Expand 15 Items
Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These centrifugal devices can quickly concentrate up to 20 ml of biological sample without valuable sample loss.
Expand 19 Items
Nanosep, Nanosep MF and NAB Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Simple, reliable processing samples of 50 to 500 μl.
Expand 26 Items
NAB Nanosep® Centrifugal Devices for RNA and DNA Extraction, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
NAB Nanosep® Centrifugal Devices for RNA and DNA Extraction, Cytiva (Formerly Pall Lab)
Expand 3 Items
National™ Centrifugal Filters, Non sterile, 2 ml, Thermo Scientific
Supplier: Thermo Scientific
Filter volumes up to 2 ml with these National™ centrifugal filters.
Expand 8 Items
EZ Double Cytofunnel™, Epredia
Supplier: Epredia
Double the number of sample presentations per run with the Epredia™ EZ Double Cytofunnel™.
Expand 2 Items
EZ Single Cytofunnel™ with Brown Filter Cards, Epredia
Supplier: Epredia
Use the Epredia™ EZ Single Cytofunnel™ with Brown Filter Cards for sample volumes of 0.4 ml or less.
Expand 1 Items
Costar® Spin-X® Centrifuge Tube Filters, Corning
Supplier: Corning
Spin-X® centrifuge tube filters use centrifugation to remove bacteria, particles, or cells from liquids, HPLC sample preparation and DNA removal from agarose or acrylamide gels.
Expand 7 Items
National™ Micro-Centrifugal Filters, Nonsterile, Thermo Scientific
Supplier: Thermo Scientific
Filter volumes as low as 50 µl with low hold-up volume with these National™ micro-centrifugal filters.
Expand 10 Items
Corning® Spin-X® UF Concentrators, Corning
Supplier: Corning
These non sterile centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.
Expand 15 Items
National™ Centrifugal Filters, Non sterile,25 ml, Thermo Scientific
Supplier: Thermo Scientific
Filter volumes up to 25 ml with these National™ 25 ml centrifugal filters.
Expand 6 Items
EZ Megafunnel, with Caps and Cytoslides™, Epredia
Supplier: Epredia
Prepare larger volume samples conveniently with the Epredia™ EZ Megafunnel with caps and slides.
Expand 1 Items
Pierce™ Protein Concentrators, Thermo Scientific
Supplier: Thermo Scientific
Protein Concentrators PES are disposable ultrafiltration centrifugal devices with a polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples such as proteins and nucleic acids.
Expand 4 Items
Protein A HP SpinTrap Spin Columns, Cytiva
Supplier: Cytiva
Protein A HP SpinTrap™ columns are designed for the enrichment of proteins of interest, immunoprecipitation and the purification of monoclonal and polyclonal antibodies from cell lysates and biological fluids.
Expand 1 Items
EZ Single Cytofunnel™ with White Filter Cards and Caps, Epredia
Supplier: Epredia
Optimize cell yield with the Epredia™ EZ Single Cytofunnel™ with white filter cards and caps.
Expand 2 Items
Spin Concentrators, 50K MWCo, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Spin concentrators are devices used to concentrate protein samples via low-speed centrifugation through MWCo filters into a collection tube.
Expand 1 Items
Vivaspin® 2 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin® concentrators are disposable ultrafiltration devices for the concentration of biological samples of 0.4 to 2 ml.
Expand 31 Items
Vivaspin Sample Concentrators, Cytiva
Supplier: Cytiva
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
Expand 24 Items
Sartorius Vivaspin® Turbo 15
Supplier: Sartorius
The Vivaspin® Turbo 15 can handle up to 15 ml sample volume in swing bucket rotors and 11 ml in fixed angle rotors accepting 50 ml centrifuge tubes.
Expand 12 Items
Vivaspin® 500 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin® centrifugal concentrators are disposable ultrafiltration devices for the concentration of biological samples. Vivaspin® 500 centrifugal filter units offer a simple, one step procedure for sample concentration with volumes from 5 to 500 µl. They can effectively be used in fixed angle rotors accepting 2.2 ml centrifuge tubes.
Expand 18 Items
Mini High-Speed Plasmid Columns, IBI Scientific
Supplier: IBI Scientific
The columns and collection tubes can be used with extra reagents from previously purchased full kits.
Expand 3 Items
Vivaspin® 6 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin® 6 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin® 6 can process 6 ml in either swing bucket or fixed angle rotors accepting standard 15 ml conical bottom test tubes.
Expand 18 Items
Vivacon® 500, Centrifugal Concentrators, Sartorius
Supplier: Sartorius
Vivacon® 500 centrifugal concentrators have been designed for DNA and protein concentration and rebuffering. They feature a horizontal membrane design that is preferred by some users for reasons of consistency with previously used procedures and reproducibility.
Expand 12 Items
Vivaspin® Endotest, Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
For 2 to 15 ml, cellulose triacetate membrane. Disposable ultrafiltration device for endotoxin concentration and removal of substances interfering with the LAL test from liquid samples. The device is a spin column based ultrafiltration unit. It can effectively be used in swing bucket or fixed angle rotors accepting 50 ml conical bottom centrifuge tubes.
Expand 1 Items
Vivaflow® 50R Ultrafiltration Units, Sartorius
Supplier: Sartorius
These are for 0.1 to 1000 ml, Hydrosart® membrane. Vivaflow 50R is a ready to use and reusable laboratory crossflow cassette for applications like cell culture supernatant concentration, virus concentration and water sample concentration. The thin channel, flip flow recirculation path of the modules provides high crossflow velocities with minimum pump speed requirements. A single module with 50 cm² active membrane area typically concentrates 250 ml by a factor of 20 in less than 30 min.
Expand 4 Items
Vivaspin® Turbo 4 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin® Turbo 4 can handle 0.04 to 4 ml sample volume in swing-out and fixed angle rotors accepting 15 ml centrifuge tubes.
Expand 12 Items
Vivaspin® 15R Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin® 15R is targeting the volume segment of 4 to 15 ml with the Hydrosart® regenerated cellulose membrane. This membrane is ideal when extremely high recovery rate and very low adsorption is required, for example in applications such as concentration and desalting of immunoglobulin fractions.
Expand 8 Items
Vivaspin® 20 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin® 20 centrifugal concentrators offer increased volume flexibility and performance to the Vivaspin® potfolio. Vivaspin® 20 handles 5 to 20 ml in swing bucket rotors and 14 ml in fixed angle rotors accepting 50 ml centrifuge tubes. Vivaspin® 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows monitoring the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate.