You searched for: Sample Bags
Sterile sample bags are ideal for transporting and storing solids, semi-solids, and liquids. Successfully protecting contents, the sealing bags permit tests to be accurately completed without worrying about possible contamination. These polyethylene bags come in a variety of thickness sizes and are ideal for packaging applications requiring ISO Class 5 cleanroom conditions. The durable fabrication of the bags resists moisture, acids, alkali, embalming fluids, and most solvents and preservatives.
VWR® Sample Sak™ Sample Bags
Supplier: VWR International
Ideal collection/transport containers for powder and granular samples including grain, seeds, feeds, coffee, powdered soaps, and even damp soil.
Expand 2 Items
Ward's® Sterile Sampling Bags
Supplier: VWR International
Gamma sterilized bags make sample collection quick and easy.
Expand 6 Items
VWR® Sterile Sample Bags
Supplier: VWR International
These sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
Expand 39 Items
VWR® Sterile Opaque Sample Bags
Supplier: VWR International
Made of black-pigmented resin, sterile bags protect photosensitive samples such as vitamins, chemicals, pharmaceuticals, food supplements, plant extracts, and biological tissues.
Expand 1 Items
VWR® Pink Anti-Static Bags
Supplier: VWR International
This premium anti-static bag won't corrode polycarbonate.
Expand 1 Items
VWR® Reclosable Chemotherapy Bags
Supplier: VWR International
Sturdy, durable 4 mm thick bags are ideal for transporting, storing, and disposing of items used in chemotherapy prep or administration.
Expand 2 Items
VWR® Conductive Bags, Black
Supplier: VWR International
Constructed of heavy-duty 4 mm black carbon-impregnated low density polyethylene (LDPE) film to create an extra strong static barrier.
Expand 1 Items
VWR® Blue Twist-Seal Bags
Supplier: VWR International
These blue twist-seal printed bags with safety tab and 2 round wires are made of virgin PE and have been specifically designed for the food industry.
Expand 1 Items
VWR® Sterile Sample Bags with Specimen Sponge
Supplier: VWR International
Sterile sample bags are ideal for surface sampling in a wide range of applications.
Expand 1 Items
VWR® Biopsy Bags
Supplier: VWR International
VWR® biopsy bags are specifically designed to ensure easy and effortless opening during the embedding process. Available in multiple sizes in either standard or premium grade.
Expand 4 Items
VWR® Reclosable Pink Anti-Static Bags
Supplier: VWR International
These flexible and electrostatic-free 4 mm bags qualify as Type II entitled barrier material under MIL-B-81750b.
Expand 5 Items
VWR® Water Sampling Kits
Supplier: VWR International
These sterile sampling bags are ideal for collecting chlorinated water specimens.
Expand 2 Items
Specimen Transport Bag with Requistion Pouch, Therapak®
Supplier: Therapak, LLC
These 6×9" polyethylene specimen bags feature a removable, adhesive release liner that secures specimen containers in a liquid-tight primary pouch.
Expand 1 Items
Zip Style Reclosable Transport Bags, Two-Pouch with BioHazard Symbol, Therapak®
Supplier: Therapak, LLC
Therapak® zip style bags are clear and feature a biohazard symbol imprinted on the outside.
Expand 6 Items
Science Kit Polyethylene Specimen Bags
Supplier: Avantor
Find a multitude of uses for these bags in the laboratory and field.
Expand 2 Items
Aqui-Pak™ Absorbent Pouches, Therapak®
Supplier: Therapak, LLC
Aqui-Pak™ was designed for the safe transport of laboratory specimen tubes by keeping the tubes separate and providing cushioning at the same time.
Expand 4 Items
Sampling Bags, SteriBag Blue, Bürkle
Supplier: BURKLE INC
The sterile sampling bag SteriBag Blue is especially well suited for use in the food, animal feed and pharmaceutical industries. To prevent the danger of foreign matter in production, the sample bags and the tear-off closure are colored blue. This shade of blue does not occur in a natural form in foodstuffs. Because of this it is highly visible and can be reliably discovered in a visual check if a bag or parts of it are ever mixed into production.
Expand 2 Items
Speci-Gard® Adhesive Closure Specimen Bags
Supplier: Minigrip
Speci-Gard® bags feature an innovative, liquid-tight, Press and Close® adhesive closure system, with no 'glove-to-adhesive' contact.
Expand 9 Items
Tamper-Evident Transport Bags, Elkay Plastics
Supplier: Elkay Plastics
These tamper-evident transport bags use a strong adhesive seal closure and have an integrated perforated tear line for easy opening.
Expand 6 Items
Lab Seal™ Tamper-Evident Specimen Bag, Elkay Plastics
Supplier: Elkay Plastics
Elkay’s Lab-Seal™ Tamper-Evident specimen bags with Seal-N-Rip™ removable biohazard symbol provide an means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs.
Expand 18 Items
Speci-Gard® Adhesive Closure Specimen Bag
Supplier: Minigrip
Biohazard bag with Press and Close® adhesive closure features liquid tight seal and tear line opening.
Expand 3 Items
Nalgene® Sample Bags, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Transparent LDPE sample bags with strong, 4 mil walls are ideal for collecting, storing, sealing and protecting specimens, labware and small parts.
Expand 5 Items
Lab Loc® Specimen Bags, Elkay Plastics
Supplier: Elkay Plastics
Elkay’s Lab-Loc® reclosable seal top specimen bags with Seal-N-Rip™ removable biohazard symbols provide an means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs.
Expand 13 Items
Lab-Loc® Specimen Transfer Bag, Elkay Plastics
Supplier: Elkay Plastics
Elkay’s specimen bags provide a means for transferring specimens in hospital and medical environments.
Expand 33 Items
arium® Bag, Sartorius Corp.
Supplier: Sartorius
The most innovative bagtank system.
Expand 1 Items
Speci-Cip® Reclosable Specimen Bags
Supplier: Minigrip
Bags are designed with an information pocket, a temperature indicator and tear lines.
Expand 4 Items
Speci-Gard® Specimen Transport Bag with Document Pouch
Supplier: Minigrip
Double pocket bag with press and Close® liquid-tight closure.
Expand 1 Items
SAFEGUARD® Specimen Bags
Supplier: Minigrip
Reclosable three-wall specimen transport bags feature distinct black print on an orange background, with biohazard logo and check boxes labeled Freeze, Refrigerate, and Room Temperature.
Expand 3 Items
Bag, Sterile, Whirl-Pak with Dechlorinating Agent, 177 mL, Hach
Supplier: Hach
Specially designed for chlorinated water samples (Complies with USEPA and Standard Methods criteria).
Expand 1 Items
Epredia™ Blue Nylon Biopsy Bags, Epredia
Supplier: Thermo Scientific
Ensure unrestricted fluid movement around tissue with Epredia™ Blue Nylon biopsy bags.