You searched for: Membrane Filter Capsules
Designed for fast processing and final filtration of aqueous samples, capsule filters provide efficient processing and enhanced throughput of difficult-to-filter solutions via an in-line installation. Applications include large-volume media preparation, pilot-scale manufacturing, and sterile filtration of buffers or biological solutions. These capsules are suitable for point-of-use filtration for laboratory water. Capsules are sterilized by gamma irradiation prior to bagging and the polycarbonate/polypropylene housing in most models ensure easy, rapid installation with no maintenance required.
Supracap™ 50 Depth Filter Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Supracap 50 depth filter capsules are ideal prefilters made with Seitz® high performance depth filter media for the removal of unwanted solids from solutions in laboratory and process development applications.
Expand 13 Items
Capsules with Versapor® Membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Membrane capsule is designed to filter liquids not requiring sterilization, or as a prefilter before final sterilization.
Expand 2 Items
Supracap™ 100 Depth Filter Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Supracap™ 100 depth filter capsules are ideal prefilters made with Seitz® high performance depth filter media for the removal of unwanted solids from solutions in laboratory and process development applications.
Expand 42 Items
Pleated Capsule Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Designed both for prefiltration of particulate liquids (such as serum) prior to sterilizing filtration, and removal of debris and particles from liquids not requiring sterilization.
Expand 2 Items
Mini Profile™ Capsule Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Designed for small volume production, scale-up evaluation, clarification of biological products, and gel removal.
Expand 5 Items
AcroPak™ 20 Filtration Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroPak™ 20 capsule filters are designed to efficiently process up to 2 litres of chemical and biological fluids. These filters are available in three different membrane types.
Expand 6 Items
Culture Capsules, 0.2 µm, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Specifically designed for sterilizing cell culture media.
Expand 1 Items
AcroPak™ 300 Capsule with PTFE Membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These 100% integrity-tested filters feature high flow rate venting and aggressive solvent filtration.
Expand 2 Items
AcroPak™ Filtration Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
A filter for every step in your process. From drug discovery and basic research to process development and production, Pall Laboratory’s AcroPak capsules come in a variety of membrane types and volume sizes.
Expand 24 Items
HEPA Capsule, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Designed to provide bacteria-free air when required for sterile applications.
Expand 1 Items
Kleenpak™ Capsules with HDC® II Membrane for Liquid Applications
Supplier: Cytiva
The HDC II filter medium is comprised of a high performance pleated cartridge with a polypropylene filter medium.
Expand 2 Items
Mini Capsule Filter, 0.45 µm, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Use both for prefiltration of particulate liquids prior to final sterilizing filtration, and for removal of particles and debris from liquids not requiring final sterilization.
Expand 1 Items
GWV High-Capacity Groundwater Sampling Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These disposable filters are designed specifically for the filtration of groundwater prior to dissolved metals analysis.
Expand 3 Items
Carbon Capsule Filter, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Removes odor, colloids, and organics from liquids using activated carbon.
Expand 1 Items
Envirochek™ and Envirochek™ HV Sampling Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Designed for collection and recovery of waterborne contaminants, Cryptosporidium, and Giardia, with typically greater than 70% recovery of target organisms.
Expand 2 Items
AquaPrep™ and AquaPrep™ 600 Groundwater Sampling Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Disposable filters designed specifically for the filtration of groundwater prior to dissolved metals analysis.
Expand 2 Items
Whatman™ Polycap™ AS Disposable Filter Capsules, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman Polycap AS (Aqueous Solution) combines a nylon membrane filter and glass microfiber (GMF) prefilter for a long lifetime device that can filter large volumes of even difficult, particulate-laden samples.
Expand 6 Items
Whatman™ Polycap™ TF Disposable Filter Capsules, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman Polycap TF filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a polypropylene housing for use with organic solvents and chemically aggressive solutions.
Expand 24 Items
Whatman™ Polycap™ TC Disposable Filter Capsules
Supplier: Cytiva
Whatman Polycap TC filtration capsules from Cytiva feature dual layer polyethersulfone (PES) membranes and provide efficient filtration for aqueous solutions.
Expand 20 Items
Whatman™ Polycap™ HD Disposable Filter Capsules
Supplier: Cytiva
Polycap HD (Heavy Duty) capsule filter offers a broad range of particle size retention ratings and excellent filtrate purity with polypropylene filters (PP).
Expand 26 Items
Whatman™ Polycap™ SPF Filter Capsules, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman Polycap SPF (Serum Prefilter) Capsule Filters are optimized for prefiltration applications. They are typically used upstream of a Polycap AS or a Polycap TC capsule during enzyme and buffer filtration.
Expand 2 Items
Whatman™ Polycap™ GW, Ground Water Sampling Capsule Filter, Whatman products (Cytiva)
Supplier: Cytiva
Specifically designed for field sampling, Cytiva's Whatman Polycap GW ground water sampling filter capsules are a polyethersulfone (PES) filter that can be used as a convenient in-line filter unit.
Expand 2 Items
PROPOR SG MURUS Capsules, Parker Bioscience
Supplier: PARKER HANNIFIN TE
PROPOR SG sterilizing grade filters feature a microbially retentive polyethersulfone membrane for fast, reliable and cost-effective sterile filtration of pharmaceutical fluids.
Expand 24 Items
Sartolab® P20 and P20 Plus Pressure Filtration Units, Sartorius
Supplier: Sartorius
Ready to use units which, attached to a membrane pump or tube pump, quickly and reliably sterilise 100 ml to 5 litres of media and aqueous solutions.
Expand 3 Items
Dissolved Oxygen Cap Membrane Kit, YSI
Supplier: YSI
Membranes are 1 ml FEP fluoropolymer film. Response time (T95 - from 0 to 100%) is 18 seconds.
Expand 1 Items
PROPOR HC MURUS Capsules, Parker Bioscience
Supplier: PARKER HANNIFIN TE
PROPOR HC sterilizing grade filters have been specifically designed for the effective and economical processing of difficult to filter solutions.
Expand 8 Items
Parker® PROPOR SG DEMI Capsules
Supplier: PARKER HANNIFIN TE
PROPOR SG sterilizing grade filters feature a microbially retentive polyethersulphone membrane for fast, reliable and cost-effective sterile filtration of pharmaceutical fluids. Silicone seals material.
Expand 18 Items
TETPOR AIR DEMI Capsules
Supplier: PARKER HANNIFIN TE
TETPOR AIR sterilization filter capsules offer exceptional filtration performance while providing the highest levels of biosecurity throughout the process industry.
Expand 9 Items
Parker® PROPOR HC DEMI Capsules
Supplier: PARKER HANNIFIN TE
PROPOR HC sterilizing grade filters have been specifically designed for the effective and economical processing of difficult to filter solutions.
Expand 18 Items
Kleenpak™ Capsules with HDC® II Membrane for Liquid Applications
Supplier: Cytiva
The HDC II filter medium is comprised of a high performance pleated cartridge with a polypropylene filter medium.