You searched for: Jars
Jars are offered in different sizes and shapes to accommodate any sample or procedure needed. The containers are manufactured using opaque or clear materials for optimum visual monitoring and measurements. Jars may be purchased with a cap for the wide mouth openings to seal substances securely. With autoclaving ability, select vessel options allow sterilization procedures for economical reusing without contaminating experiments. From general to the most specific use, jars are designed to achieve any task.
VWR® Staining Jar with Cover, Hellendhal Type
Supplier: VWR International
This tapered jar is made of optically clear PMP (polymethylpentene).
Expand 1 Items
VWR® TraceClean® Straight Side Glass Jars, Clear
Supplier: VWR International
These TraceClean® glass jars are used primarily for the collection of soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
Expand 9 Items
VWR® Tall Straight Sided Jars, Clear
Supplier: VWR International
VWR® Clear Tall Straight Sided Jars are manufactured from soda-lime glass that conforms to USP Type III requirements.
Expand 7 Items
VWR® TraceClean® Straight-Sided Wide Mouth Jars, Glass
Supplier: VWR International
Type III (soda-lime) glass jars are for collection of soil, sediment, and slurry matrices.
Expand 22 Items
VWR® TraceClean® Wide Mouth Jars, Glass
Supplier: VWR International
Type III (soda-lime) jars are for collection of solid matrices or liquids dispensed from bailers or peristaltic tubing.
Expand 18 Items
VWR® Straight Sided Jars, Clear and Amber
Supplier: VWR International
VWR® Straight Sided Jars are manufactured from soda-lime glass that conforms to USP Type III requirements.
Expand 18 Items
VWR® Jars, PTFE
Supplier: VWR International
Isostatically molded, pure FDA approved PTFE molded stackable jars have thick wall construction and a smooth internal finish.
Expand 8 Items
VWR® TraceClean® Mayo Tall Glass Jars, Clear
Supplier: VWR International
These TraceClean® glass jars are used primarily for the collection of soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
Expand 5 Items
VWR® TraceClean® Boston Round Glass Jars, Narrow Mouth, Amber
Supplier: VWR International
These TraceClean® glass jars are used primarily for the collection of aqueous media for testing semi-volatiles, pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
Expand 4 Items
VWR® TraceClean® Wide Mouth Jars, HDPE
Supplier: VWR International
Straight-sided jars are for collecting clays, slurries, and sediment matrices for use in geotechnical, environmental. Toxicity Characteristic Leaching Procedure (TCLP) metals, inorganic compounds, and benthic organism studies.
Expand 9 Items
VWR® TraceClean® Straight Sided Containers, HDPE
Supplier: VWR International
These TraceClean® containers are used primarily for the collection of soils, clays, slurries, and sediment media for geotechnical testing, Toxicity Characteristic Leaching Procedure (TCLP), metals, total metals, inorganic analytes, and benthic organism studies.
Expand 3 Items
Ward's® Killing Jars
Supplier: Avantor
The improved design of these jars incorporates a bottom pad that better absorbs killing agent, so that fumes are contained and insects are knocked down quickly, causing less damage to specimens.
Expand 2 Items
Charger for Killing Jar
Supplier: Ward's Science
Includes Squeeze Bottle for Convenient Application
Expand 1 Items
Nalgene® Polycarbonate Multipurpose Jars with Covers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Nalgene® Polycarbonate Multipurpose Jars are useful as water baths, aquaria, terraria or freeze-drying chambers
Expand 3 Items
SP Bel-Art Mason Jars, Wide Mouth, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Highly shock-resistant polypropylene, heavy-walled Mason Jars are ideal for industrial waste sampling, long-term storage, and a variety of laboratory uses.
Expand 5 Items
Straight Side Glass Bottles, Clear, C&G Containers
Supplier: C&G Containers
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics
Expand 22 Items
Wide Mouth Glass Jars, Clear, C&G Containers
Supplier: C&G Containers
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics
Expand 3 Items
Nalgene® Polypropylene Mason Jars, Thermo Scientific
Supplier: Thermo Fisher Scientific
Durable, break-resistant polypropylene jars are autoclavable
Expand 4 Items
Anaerobe Jar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
This anaerobic jar is specially formulated for temperature stability and durability
Expand 1 Items
Nalgene® Polypropylene Jars with Cover, Thermo Scientific
Supplier: Thermo Fisher Scientific
For storing and transporting solids
Expand 2 Items
Clear Glass Straight Sided Jars
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Soda lime glass jars feature polypropylene screw caps and polyvinyl liners.
Expand 3 Items
Nalgene® Polycarbonate Straight-Sided Wide-Mouth Jars, Thermo Scientific
Supplier: Thermo Fisher Scientific
Excellent for cold room and refrigerated use
Expand 7 Items
Straight-Sided Jars, Clear, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Wheaton Straight Sided jars are manufactured from clear soda lime glass that conforms to USP Type III requirements.
Expand 17 Items
Wide Mouth Glass Packer Bottles, Amber, C&G Containers
Supplier: C&G Containers
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles (Light Sensitive compounds), Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics
Expand 21 Items
Nalgene® Polypropylene Straight-Sided Jars, Thermo Scientific
Supplier: Thermo Fisher Scientific
For storing and collecting lab supplies and specimens
Expand 7 Items
Nalgene® Vacuum Chambers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Vacuum chamber consists of transparent jar, neoprene gasket and vacuum plate with adapter for 1/4" I.D tubing.
Expand 3 Items
EP Scientific, Depyrogenated Jars, with Closure, Thermo Scientific
Supplier: Thermo Scientific
Thermo Scientific™ Depyrogenated Glass Containers are specially prepared to meet endotoxin levels of less than 0.06 EU/mL for use in terminal sterilization, water sampling and product storage.