You searched for: Eyewash Stations and Eyewash Bottles
Eyewash stations for quick and accessible care in emergencies are offered at VWR. Open, wall mounted models feature bottles of sterile saline with twist-off tops and flexible eye cups; select closed stations for dirt and dust prone work spaces. Delivery systems offer an automated, gravity-fed and hands-free solution without plumbing. Convert your lab faucet to an eyewash station with a faucet mount unit that attaches with a threaded inlet. Plumbed eyewash stations that require a dedicated water source are also available.
SP Bel-Art Eye Wash Station, Bel-Art Products, a part of SP
Supplier: Bel-Art
This easily recognized, ready-to-use eye wash station comes complete with eye wash bottle, dust cover, mirror, and instructions for use and service.
Expand 2 Items
Hughes Portable, Self-Contained 16 Gallon Gravity-Fed Eyewash Station, Justrite®
Supplier: Justrite
The hughes portable, self-contained 16 gal. gravity-fed eyewash station requires no plumbing, and is suitable for use wherever continuous water is not available.
Expand 1 Items
Hydrosep® Eye Wash Bottle, Encon
Supplier: Encon Safety Products
Recommended cleaning and fluid replacement every 90 to 120 days in unsealed, gravity-fed eyewashes.
Expand 1 Items
Eyesaver® Eye/Face Wash Station, Speakman®
Supplier: Speakman
These wall-mounted eye/face washes feature dual spray heads activated by a stainless steel push handle.
Expand 2 Items
Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety Products
This bottle contains a sterile, buffered saline solution for emergency eye care.
Expand 3 Items
Eyesaver Drench Hose and Eyewash Combination, Model SE-925, Speakman®
Supplier: Speakman
Speakman® Drench hose and eyewash combinations provide great flexibility in laboratories and other industrial areas for flushing the eyes and body.
Expand 1 Items
Single Bottle Eye Wash Station, Brady®
Supplier: Brady Worldwide
Our single bottle eye wash stations feature a drop down front panel that allows quick access to your eyewash bottles (not included).
Expand 1 Items
GravityFlo® Portable Eyewash, SE-4400, Speakman®
Supplier: Speakman
The SE-4400 portable eyewash allows to locate an eyewash anywhere that it is needed, even where potable water is not available.
Expand 1 Items
Hughes Gravity Fed Eyewash Insulated and Heated Jackets, Justrite®
Supplier: Justrite
Jacket, insulated, Retrofittable, perfect accessory to keep the OptiWash eyewash station a tepid temperature. Constructed of heavyweight PVC cloth and double stitched to provide extra durability, offers frost protection and reduces the effects of solar radiation, Size: 10gal
Expand 4 Items
Eyewash with Drench Hose, Speakman®
Supplier: Speakman
The eyewash features a stay-open valve and twin aerated sprays with flip-top dust caps and automatic flow control.
Expand 1 Items
Eyesaver® Deck Plate Faucet and Eyewash Combination, Speakman®
Supplier: Speakman
Eyesaver® faucet combines a sensor activated 8" deck plate gooseneck faucet with an independently operated pull-activated eyewash.
Expand 2 Items
Portable Eyewash, Speakman®
Supplier: Speakman
Perfect for environmental field labs, this portable eyewash and drench hose can be used where permanent water supply is not available.
Expand 1 Items
Lead-Free, Non-Aerated Eyesaver Faucet, Model SEF-1800NA, Speakman®
Supplier: Speakman
Speakman® Eyesaver® combination eyewashes and lavatory faucets are a revolutionary and exclusive product with a unique design.
Expand 1 Items
Hughes Portable, Self-Contained, 10 gal. Gravity-Fed Eyewash Station, Justrite®
Supplier: Justrite
Portable design with carrying handles and concealed wall mounting bracket make moving and relocating easy.
Expand 2 Items
Bradley Pressurized Portable Eye Wash
Supplier: EMEDCO
Easily transportable for fast, efficient treatment.
Expand 1 Items
Eyesaver® Multi-Spray Eye/Face Washes, Speakman®
Supplier: Speakman
Eye/face washes provide six aerated sprays for gentle yet thorough rinsing of the eyes and face.
Expand 2 Items
Optimus™ Wall Mounted Eye/Face Wash System, Speakman®
Supplier: Speakman
Wall mounted eye/face wash system equipped with a dual aerated eyewash to deliver soothing relief.
Expand 2 Items
Hughes ™ Eyewash Additive Solution, Justrite®
Supplier: Justrite
Eyewash additive preserves water in portable, self-contained eyewash stations to prevent bacterial growth.
Expand 1 Items
GRAVITYFLO® Gravity-Fed Eye/Face Wash, Speakman®
Supplier: Speakman
This portable, gravity-fed eye/face wash is perfect for environmental site locations as well as areas that are not plumbed.
Expand 1 Items
Eyesaver® Deck-Mount Swing-Out Eyewash, Speakman®
Supplier: Speakman
Low-profile counter-top eyewash unit for installations where space is at a premium.
Expand 1 Items
Fendall® Saline Eyewash Wall Stations, Honeywell Safety
Supplier: Honeywell Safety Products
Available in 470 ml (16 oz.) and 950 ml (32 oz.) single and 950 ml (32 oz.) double wall stations.
Expand 2 Items
Eyesaver® Bar Activated Eyewash Stations, Speakman®
Supplier: SPEAKMAN COMPANY SE
Wall mounted rectangular eyewash bowl.
Expand 2 Items
Eyesaver® Eyewash Station, Speakman®
Supplier: Speakman
For use in areas where workers could be exposed to harmful contaminants.
Expand 5 Items
First Aid Central Eyewash Products, Acme United
Supplier: ACME UNITED LIMITED CA
Quality eyewash stations, solutions, and kits for treating eye injuries.
Expand 13 Items
Laboratory Eyewash Station
Supplier: Bel-Art
Quick and Convenient in Case of Emergency.
Expand 3 Items
Eyesaver® Eyewash Faucets, Speakman®
Supplier: Speakman
Dedicated water ways allow the faucet and eyewash to work independently of each other.
Expand 4 Items
Drench Hose Attachment, Speakman®
Supplier: Speakman
The drench hose attachment is designed for use with the Speakman® portable GravityFlo® units.
Expand 1 Items
Faucet-Mount Eyewashes, WaterSaver Faucet
Supplier: WaterSaver Faucet
Constructed of chrome-plated brass, these faucet-mount units convert any laboratory faucet into an emergency eyewash station without special plumbing or interference with normal faucet operation.
Expand 2 Items
Safety Shower and Eye/Face Wash, Speakman®
Supplier: SPEAKMAN COMPANY SE
Combination shower with rectangular eye/face wash bowl.
Expand 3 Items
Tabletop Mounted Eyewash, Speakman®
Supplier: SPEAKMAN COMPANY SE
Twin aerated sprayheads conveniently located on the counter.