L-Methionine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
CAS Number:63-68-3
:
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063097
:149.21 g/mol
Merck Index:13,06004
Boiling Pt:278 °C (1013 hPa)
Melting Pt:136 °C
Density:1.373 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:200-562-9
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (C₅H₁₁NO₂S) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):22.4 - 24.7 Degree
- USP - pH:5.6 - 6.1
- USP - Loss on Drying at 105°C:≤0.3 %
- USP - Residue on Ignition:≤0.4 %
- USP - Chloride (Cl):≤0.05 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Iron (Fe):≤0.003 %
- USP - Related Compounds - Methionine sulfoxide:≤0.1 %
- USP - Related Compounds - N-Acetyl-D,L- methionine:≤0.2 %
- USP - Related Compounds - Any unspecified impurity:≤0.1 %
- USP - Related Compounds - Total unspecified impurities:≤0.30 %
- EP/BP - Assay (C₅H₁₁NO₂S) (dried basis):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:5.5 - 6.5
- EP/BP - Specific Rotation [Å]²º^D (+):22.5 - 24.0 Degree
- EP/BP - Related Substances: Unspecified Impurities, each:≤0.10 %
- EP/BP - Related Substances: Total Impurities:≤0.3 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (C₅H₁₁NO₂S) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):21.0 - 25.0 Degree
- JP - pH:5.2 - 6.2
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.021 %
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.30 %
- JP - Residue after Ignition:≤0.10 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2085 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR2085
SDS
spec_JT2085-07 spec_JT2085-5
Product specification
Frequently Bought Together