Order Entry
Canada
Orders LinkContactUsLinkComponent
L(+)-Histidine USP, Multi-Compendial

L(+)-Histidine USP, Multi-Compendial

L(+)-Histidine USP, Multi-Compendial
MOC:  Regulated
Certificates
L(+)-Histidine USP, Multi-Compendial
MOC:  Regulated

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

White Crystalline solid.

Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1.44 g/cm³ (20 °C)
  •  :Management of Change (MOC) category = R
  • Storage Temperature:Ambient
  • EINECS:200-745-3
Specifications for Product Family

Specification Test Results

  • USP - Assay (C₆H₉N₃O₂) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation [Å]²⁵ ^D (+):
    12.6 - 14.0 Degree
  • USP - pH:
    7.0 - 8.5
  • USP - Loss on Drying at 105°C:
    ≤ 0.2 %
  • USP - Residue on Ignition:
    ≤ 0.4 %
  • USP - Chloride (Cl):
    ≤ 0.05 %
  • USP - Sulfate (SO₄):
    ≤ 0.03 %
  • USP - Iron (Fe):
    ≤ 0.003 %
  • USP - Related Compounds - Individual Impurities:
    ≤ 0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤ 2.0 %
  • EP/BP - Assay (C₆H₉N₃O₂) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation [Å]²º^D (+):
    11.4 - 12.4 Degree
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤ 0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤ 0.5 %
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤ 0.02 %
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    ≤ 0.5 %
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • JP - Assay (C6H9N3O2) (dried basis):
    99.0 - 101.0 %
  • JP - Ammonium (NH₄):
    ≤ 0.02 %
  • JP - Chloride (Cl):
    ≤ 0.021 %
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Iron (Fe):
    ≤ 10 ppm
  • JP - Identification:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤ 0.30 %
  • JP- Optical Rotation [Å]²º^D (+):
    11.8 - 12.8 Degree
  • JP - Related Substances:
    Passes Test
  • JP - pH:
    7.0 - 8.5
  • JP - Residue on Ignition:
    ≤ 0.10 %
  • JP - Sulfate (SO₄):
    ≤ 0.028 %
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.