Orders LinkContactUsLinkComponent
Analyzer Sample Cups, Greiner Bio-One
Analyzer Sample Cups, Greiner Bio-One
Analyzer Sample Cups, Greiner Bio-One
Supplier:  Greiner Bio-One
Certificates
Analyzer Sample Cups, Greiner Bio-One
Analyzer Sample Cups, Greiner Bio-One
2 Options Available Below
Product Details & Documents
Sample cups are well suited for a number of established analyzer systems including Centrifichem, Coulter Counter®, Gemsaec, Hitachi®, Technicon™, and Sysmex® systems.
  • Polyethylene or polystyrene construction
  • Suited for a number of established analyzer systems
  • Clear
Sample cup 82051-154 is constructed of polyethylene; all others are constructed of polystyrene.
Documents
Product Family Options
Product Information
  • Capacity
    W×H
    Availability
    Price
  • 82051-134
    Sample Cup for Hitachi® Analyzers, Conical/Skirted
    Sample Cup for Hitachi® Analyzers, Conical/Skirted
    1.5 ml (0.05 oz.)
    17×38 mm
    Log in for availability
    Log in for availability
    Case of 5,000
    Case of 5,000 - $298.74
    $298.74
    New Customer Promotional Price
    Case of 5,000
       $298.74
    Supplier Number: 729101
  • 82050-936
    Sample Cup for Coulter Counter® Analyzers, Flat
    Sample Cup for Coulter Counter® Analyzers, Flat
    25 ml (0.85 oz.)
    32×60 mm
    Log in for availability
    Log in for availability
    Case of 1,250
    Case of 1,250 - $408.82
    $408.82
    New Customer Promotional Price
    Case of 1,250
       $408.82
    Supplier Number: 668102