EPA method 418.1 TPH Calibration Mixture, Restek
Catalog # RK30080
Supplier: Restek
CAS Number:
Specifications
- Description:EPA Method 418.1 TPH Calibration Mixture
- Cat. no.:RK30080
- Supplier No.:30080
Specifications
About this item
For the infrared spectrophotometric method for the determination of Total Recoverable Petroleum Hydrocarbons.
- Mixture contains chlorobenzene, 25.0% (v/v), n-Hexadecane, 37.5% (v/v), and 2,2,4-Trimethylpentane, 37.5% (v/v)
Mixture is packaged at the stated percentages in a 1 ml ampoule. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).