Orders LinkContactUsLinkComponent
Protector® Demonstration Hoods, Labconco®
 :   Labconco
Protector® Demonstration Hoods, Labconco®
 :  Labconco
Protector® Demonstration Hoods, Labconco®
Protector® Demonstration Hoods, Labconco®
Designed for classroom use, these lightweight hoods feature an epoxy-coated aluminum frame with tempered safety glass front sash, sides, rear baffle, and top.
Hood systems include a white, epoxy-coated tubular steel cart with a black 2.5cm (1") thick solid epoxy dished work surface, a handle bar, a bottom shelf, and 10.2 cm (4") diameter toe-locking casters. Also included is a FilterMate™ Portable Exhauster, which requires either two carbon filters or one carbon filter and one HEPA filter (filters sold separately).
The organic carbon filter cell is a type AC filter that contains 5.4 kg (12 lbs.) of granular activated carbon media suitable for organic solvents including acetone, ethyl acetate, toluene, and xylene. The formaldehyde carbon filter cell is a type C1 filter that contains 6.4 kg (14 lbs.) of impregnated carbon media suitable for vapors from formaldehyde. The ammonia filter cell is a type CX filter that contains 7.3 kg (16 lbs.) of impregnated carbon media suitable for ammonia and low-molecular weight amines. The HEPA filter cell is 99.99% efficient on particulates 0.3 µm in size; however, it should not be used to capture biohazardous materials or volatile organic material. The syringe kit is for carbon saturation detection (detector tubes sold separately).
Hoods have a nominal width of 91.4 cm (3') and an interior depth of 58.4 cm (23") to easily accommodate a variety of apparatus. Hoods feature the Clean-Sweep™ air foil that allows air to sweep the work surface for maximum containment; two utility ports with iris openings for connection of tubing, cords, and cables; and an ergonomic angled sash that reduces glare. Flexible exhaust options enable the hood to be connected to ductwork for exhausting to the outside or to the optional FilterMate™ Portable Exhauster (see 82010-632 series) for filtered removal of fumes and/or particulates.
 : All hoods conform to ASHRAE 110-1995 standards. The 115 V models are ETL and ETL-C listed. The 230 V models are CE marked.
 : Hoods 89000-162 and 89000-104 require a connection to a remote blower, house exhaust, or FilterMate™ Portable Exhauster (see 82010-632 series). They also require a work surface and a supporting base. For Guardian airflow monitors, see 82007-280 series. For exhaust dampers, remote blowers, transition adapters for connecting ductwork, and other accessories, contact your VWR sales representative.
Product Family Options
Demonstration Hoods
  • Electrical
    Depth
    Width
    Enclosure height
    Rediship
    Shipping weight
  • 584 mm (23")
    914 mm (36")
    22⁸/₁₀"
    No
    75 lbs.
    Each
    Each - $6,860.00
    $6,860.00
    Each
       $6,860.00
     : 3944300
  • 584 mm (23")
    914 mm (36")
    32"
    No
    85 lbs.
    Each
    Each - $7,770.00
    $7,770.00
    Each
       $7,770.00
     : 3944301
Demonstration Hood Systems
  • Electrical
    Depth
    Width
    Enclosure height
    Rediship
    Shipping weight
  • 115 V, 60 Hz, 10 A
    584 mm (23")
    914 mm (36")
    22⁸/₁₀"
    No
    125 lbs.
    Each
    Each - $17,730.00
    $17,730.00
    Each
       $17,730.00
     : 3945000
  • 115 V, 60 Hz, 10 A
    584 mm (23")
    914 mm (36")
    32"
    No
    135 lbs.
    Each
    Each - $18,650.00
    $18,650.00
    Each
       $18,650.00
     : 3945001
  • 230 V, 50/60 Hz, 5 A
    584 mm (23")
    914 mm (36")
    22⁸/₁₀"
    No
    125 lbs.
    Each
    Each - $18,110.00
    $18,110.00
    Each
       $18,110.00
     : 3945020
  • 230 V, 50/60 Hz, 5 A
    584 mm (23")
    914 mm (36")
    32"
    No
    135 lbs.
    Each
    Each - $19,050.00
    $19,050.00
    Each
       $19,050.00
     : 3945021
Accessories
  •  
    Description
    REDISHIP
    Shipping Weight
  • 82010-692
    Solid Epoxy Dished Work Surface for 3' Models
    Yes
    72 lbs.
    Each - $2,010.14
  •  
    Description
    REDISHIP
    Shipping Weight
  • 82010-710
    Fluorescent Light Kit, 115V, 60Hz, 10A
    No
    15 lbs.
    Each - $1,710.00
  •  
    Description
    REDISHIP
    Shipping Weight
  • 82010-712
    Fluorescent Light Kit, 230V, 50Hz, 10A
    No
    15 lbs.
    Each - $1,820.00
  •  
    Description
    REDISHIP
    Shipping Weight
  • 89002-372
    Cart, White Epoxy-Coated Tubular Steel
    No
    50 lbs.
    Each - $2,930.00
  •  
    Description
    Shipping Weight
  • 82010-732
    Syringe Kit
    0.5 kg (1 lbs.)
    Each - $85.10
  •  
    Description
    For Use With
    REDISHIP
  • 82010-684
    FilterMate™ Carbon Filter, Ammonia
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Each - $1,110.00
  •  
    Description
    For Use With
    REDISHIP
  • 82010-688
    Bag-In/Bag-Out Bag for HEPA Filter
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Each - $623.00
  •  
    Description
    For Use With
    REDISHIP
  • 82010-680
    FilterMate™ Carbon Filter, Organic
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Each - $644.00
  •  
    Description
    For Use With
    REDISHIP
  • 82010-686
    FilterMate™ HEPA Filter
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Each - $1,054.50
  •  
    Description
    Shipping Weight
  • 46000-246
    Ammonia Detector Tubes, Set of 3
    0.5 kg (1 lbs.)
    Each - $199.00
  •  
    Description
    For Use With
    REDISHIP
  • 82010-682
    FilterMate™ Carbon Filter, Formaldehyde
    Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes
    No
    Each - $1,110.00
  •  
    Description
    Shipping Weight
  • 46000-250
    Formaldehyde Detector Tubes, Set of 3
    0.5 kg (1 lbs.)
    Each - $244.00