PYREX® Burets, Class B, Straight Bore Fluoropolymer Resin Stopcock, Colored Scale, Corning
Supplier: Corning
Certificates
About this item
Borosilicate glass.
- Microfinish of the barrel provides a precision fit
- Durable colored markings
Borosilicate glass.
These burets are suitable for use in school and institutional laboratories where the accuracy of class A models is not required.
The tips are carefully drawn for accuracy to insure proper drainage rate.
Delivery: Burets are supplied with a 2 mm PTFE stopcock plug and dust cover.
Specifications
- Catalog No:
- 17452-142
- 17452-164
- 17452-109
- 17452-120
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey