PYREX® Burets, Class A, Colored Graduations, Fluoropolymer Resin Stopcock, Corning
Supplier: Corning
Certificates
About this item
Borosilicate glass, class A.
- Red colored graduations feature permanent markings, fine clear lines
- Large easy to read numbers
- Smooth micro-finish stopcock assures leak-resistant performance
Burette tips are carefully drawn from precision bore tubing to ensure proper drainage rates.
Certifications: Capacity tolerance is established by ASTM E287 and calibrated in accordance with ASTM E542.
Delivery: Burets are shipped complete with PTFE stopcock and dust cover.
Specifications
- Catalog No:
- 17452-426
- 17452-368
- 17452-404
- 17452-380
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey