Specifications
- Temperature range:–50 to 150°C (–58 to 302°F)
- Description:Digital Thermometer with Probe
- Temperature accuracy:±0.05°C
- Temperature resolution:0.001°
- W×D×H:89×140×32 mm (31/2 × 51/2 × 11/4")
- Probe:3.2 dia. x 159L mm (1/8 x 61/4")
- Cat. no.:61220-601
Specifications
About this item
Ideal for measuring temperatures in cuvettes, gas systems, reagents, and chemical solutions, or for routine measurements.
- Traceable®
- Read °C and °F
- Compact and portable
- Resolution of 0.001°
High-contrast LCD features 12.7mm (1/2) digits
Units Read °C or °F at the touch of a button.
The Data Acquisition System captures, displays, and stores readings in Windows®. Data can be read as-is or imported into databases, spreadsheets, word processors, and statistical programs.
Readings are updated three times a second. Features include hold button, minimum/maximum memories, and 50 data logger memories. Units operate on 9V alkaline battery (included) or optional 115V adapter/recharger.Thermometer 61220-601 comes with a standard quick-response, stainless steel triple purpose probe that features a 1.5m (5') cable and can be used with liquids, semi-solids, or air/gas.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
For improved performance, probes should be calibrated as a system with the meter.