About this item
For safe, easy, effective spill treatment in the laboratory
- Agents are formulated to rapidly clean up chemical spills and vapors
- Each bottle has a convenient dispensing cap for shaking or pouring
Caustics and acids are converted into nonhazardous compounds that meet EPA specifications for nonhazardous waste, simplifying cleanup and disposal. Solvent spills can be easily treated and transported with the absorbent in the solvent spill kit. This absorbent will also act as a vapor suppressant. The formaldehyde spill kit contains a two-part neutralization and immobilization system that effectively neutralizes formaldehyde solution spills. The multipurpose kit can treat up to a 4L (1.1gal.) solvent spill or a 2.5L (0.7gal.) acid and caustic spill. The mercury spill kit controls mercury spills in minutes using a simple, two-step procedure. Also included is a mercury vapor suppressant absorbent which is ideally suited to penetrate inaccessible areas such as cracks and crevices.
SpillSolv® kits are available for acid, caustic, solvent, formaldehyde, or mercury spills. Also available is a multipurpose spill kit that contains spill response materials for a combination of spills.
Each kit includes goggles, gloves, disposal bags, scoops, sponge, and guide. The mercury spill kit also includes syringes and Tyvek® booties. The formaldehyde spill kit also includes spray heads. Purchase EM-SX1300S-1 to replace the standard accessories set, and EM-SX1340S-1 to replace the formaldehyde accessories set.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberCOEMSX13001NO_11232009 EM-SX1300-1_11232009 EM-SX1310-10_11232009 EM-SX1310-1_11232009 EM-SX1320-1_11232009
SDS
Focus Safety Volume 1 Issue 1 Safety Essentials Volume 2 VWR Histology Brochure VWR® Focus Production Volume 3
Product Literature
VWR GHS Compliance Letter (US)