Axygen® PCR Tube Storage Racks, Corning
Supplier: Corning
Certificates
About this item
PP with lid.
- Fluorescent color base with translucent lid
- Durable, chemically inert, and can withstand extended duration at sub-zero temperatures
Designed to hold 0.2, and 0.5 ml PCR tubes.
Ordering information: Assorted pack includes one each of blue, green, violet, red and yellow.
Specifications
- Catalog No:
- 80086-082
- 80086-074
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa