ContactUsLinkComponent
VWR® Traceable® Waterproof Type-K Thermometer
VWR® Traceable® Waterproof Type-K Thermometer
Catalog # 89094-738
Supplier:  VWR International
undefined
VWR® Traceable® Waterproof Type-K Thermometer
Catalog # 89094-738
Supplier:  VWR International
Specifications
  • Description:
    Waterproof Thermometer
  • Temperature accuracy:
    ±0.3% of reading plus 1°C (2°F) for –50 to 1000°C (–58 to 1832°F); ±0.5% of reading plus 1°C (2°F) for All Other Temperatures
  • Temperature resolution:
    0.1° from –200 to 650°C, 0.1° from –328 to 999°F, 1° for all other temperatures
  • W×D×H:
    7.6×3.2×18.4 cm (3×11/4 × 71/4")
  • Weight:
    226.8 g (8 oz.)
  • Cat. no.:
    89094-738
  • Range:
    –200...1300 °C (–328...2372 °F)
Specifications
About this item
Wide-range thermometer has shock-resistant ABS plastic casing.
  • Traceable
  • Temperature in °C and °F
  • Waterproof design
  • Highly visible display
  • Minimum and maximum temperature recall
Waterproof design stops dirt, dust, fumes, and water, making it ideal in harsh lab or plant environments. Features include °C or °F display, minimum and maximum temperature recall, differential temperature from a set point, hold button to capture current reading, and backlight button. The microprocessor updates the display 2.5 times per second. The 1.6 cm (5/8") digits are readable from 3m (10').
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with 9-volt battery, carrying case, fast-response type-K beaded probe with 1.2 m (4') cable, and waterproof rubber cover for probe connector. For other available type-K probes (sold separately), see the 61161-372 series.