HD Super™ Mobile Worktables, Metro™
Supplier: Metropolitan Wire
Certificates
About this item
Exceptionally stable, easy-to-clean conductive tables are ideal for the cleanroom, classroom, or laboratory.
- Seamless 14-gauge Type 304 stainless steel work surface with a support structure that virtually eliminates drumming
- Fitted with four 12.7 cm (5") diameter polyurethane swivel casters, two with wheel brakes
- Work surface fits over plastic sleeves at the top of the posts
Tables are available with either a bottom shelf, a three-sided horizontal tubular frame support, or a horizontal "H" frame support. Shelves are adjustable in 5.1 cm (2") increments along the height of the posts. Tables feature 3.8 cm (15/8") diameter posts and tapered sleeves and corners to assure a rigid, sturdy structure.
Certifications: NSF listed.
Customers Who Bought This Also Bought