Orders LinkContactUsLinkComponent
Explosion-Proof Stirrer, Lightnin®
115 V, 60 Hz, 40 W, 0 - 1750 rpm, 200 L (53 gal.), ¹⁄₂₀ hp W, 13.6 kg (30 lbs.)
Catalog # 58963-303
Supplier:   Lightnin
 :  800013
Explosion-Proof Stirrer, Lightnin®
Catalog # 58963-303
Supplier:  Lightnin
Explosion-Proof Stirrer, Lightnin®
Explosion-Proof Stirrer, Lightnin®
115 V, 60 Hz, 40 W, 0 - 1750 rpm, 200 L (53 gal.), ¹⁄₂₀ hp W, 13.6 kg (30 lbs.)
Price
$21,194.87
Log in for availability
Each
Product Details & Documents
Designed for use in flammable or explosive environments, this constant speed stirrer readily mixes low-viscosity fluids or smaller amounts of more viscous liquids.
 : Stirrer must be hardwired into the user's electrical supply. Plug and cord are not included. Stirrer comes with instruction manual, bronze ring clamp, stainless steel chuck and tube, 24" (61 cm) long stainless steel accessory shaft, and one A310 stainless steel impeller with an 8.6 cm (313/32") diameter blade and 7.9 mm (5/16") diameter bore.
Specifications
  • Electrical:
    115 V, 60 Hz, 40 W
  • Shipping weight:
    13.6 kg (30 lbs.)
  • Speed range:
    0 - 1750 rpm
  • Horsepower:
    ¹⁄₂₀ hp W
  • Max. volume:
    200 L (53 gal.)
Accessories
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Impeller Blades (A100 Series)
    Stainless Steel Impeller Blades (A100 Series)
    58958-200
    7.9 mm bore Ø×2.5 cm blade Ø (5/16×1.0")
    Log in for availability
    Each - $924.32
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Impeller Blades (A200 Series)
    Stainless Steel Impeller Blades (A200 Series)
    58958-066
    7.9 mm bore Ø×5.1 cm blade Ø (5/16×2.0")
    Log in for availability
    Each - $2,867.77
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Impeller Blades (A100 Series)
    Stainless Steel Impeller Blades (A100 Series)
    58958-222
    7.9 mm bore Ø×3.8 cm blade Ø (5/16×1.5")
    Log in for availability
    Each - $835.14
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Shafts without Impeller
    Stainless Steel Shafts without Impeller
    58958-164
    30.5 cm long×9.5 mm Ø shaft (12×3/8")
    Log in for availability
    Each - $333.08
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Shafts without Impeller
    Stainless Steel Shafts without Impeller
    58958-186
    45.7 cm long×9.5 mm Ø shaft (18×3/8")
    Log in for availability
    Each - $452.70
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Impeller Blades (A100 Series)
    Stainless Steel Impeller Blades (A100 Series)
    58958-233
    7.9 mm bore Ø×6.9 cm blade Ø (5/16×2.7")
    Log in for availability
    Each - $828.09
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Impeller Blades (A100 Series)
    Stainless Steel Impeller Blades (A100 Series)
    58958-244
    7.9 mm bore Ø×5.1 cm blade Ø (5/16×2.0")
    Log in for availability
    Each - $1,021.50
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Shafts without Impeller
    Stainless Steel Shafts without Impeller
    58958-255
    30.5 cm long×7.9 mm Ø shaft (12×5/16")
    Log in for availability
    Each - $268.40
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Shafts without Impeller
    Stainless Steel Shafts without Impeller
    58958-266
    45.7 cm long×7.9 mm Ø shaft (18×5/16")
    Log in for availability
    Each - $428.24
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Stainless Steel Shafts without Impeller
    Stainless Steel Shafts without Impeller
    58958-277
    61.0 cm long×7.9 mm Ø shaft (24×5/16")
    Log in for availability
    Each - $384.33