ContactUsLinkComponent
SP Bel-Art Spinpak™ Magnetic Stirring Bar Assortment with Restrainer, Bel-Art Products, a part of SP
Catalog # 89082-458
undefined
SP Bel-Art Spinpak™ Magnetic Stirring Bar Assortment with Restrainer, Bel-Art Products, a part of SP
Catalog # 89082-458
Supplier Number:  F37158-0010
Specifications
  • Description:
    Spinpak™ Magnetic Stirring Bar Assortment with Restrainer
  • Cat. no.:
    89082-458
  • Supplier No.:
    371580010
Specifications
About this item
A 10-pack of the most commonly used Spinbar® Octagonal Magnetic Stirring Bars in a variety of colors and sizes.
  • PTFE low-friction coating
  • Durable and inert
  • Interrupted profile provides greater surface area and added turbulence
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
Eight-sided Spinbar® Octagonal Magnetic Stirring Bars provide greater surface area and added turbulence when compared to smooth cylindrical bars. An integral pivot ring on each stirring bar aids in reducing friction and chattering. Made of Alnico magnet encapsulated in FDA-grade, USP Class VI Teflon® PTFE low friction coating, they are durable and inert. Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification. Autoclavable. Included Spinbar® Restrainer fits comfortably in one hand and retains Spinbar® Magnetic Stirring Bars up to 80 mm (3”) long by holding them against vessel walls; eliminates splashing hazards when pouring. Includes Spinbar® sizes: 2 each - 1/2 x 1/8”, 1/2 x 5/16”, 1 x 5/16”, 11/2 x 5/16”, 2 x 5/16”.