Synthware Solvent Still Head with Drainage Arm, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Still features a syringe port with a [ST] 14/20 outer joint for a rubber septum and a 4 mm PTFE stopcock.
- Heavy wall Borosilicate construction
- Ground glass joint
- Teflon® stopcocks
- Ground Glass drip tip
Solvent is drained through a 2 mm T-bore PTFE stopcock. The drainage arm has a [ST] 14/20 inner joint.
Frequently Bought Together