Synthware Pressure Equalizing Funnel, Kemtech America
Supplier: Kemtech America
Jump to:
About this item
Funnels are supplied with a 1:5 PTFE stopcock.
- Heavy wall borosilicate construction
- Ground glass joint
- Teflon® or glass(G) stopcock
- Graduated
Our dropper/addition funnel is specifically designed for the controlled transfer of liquids.
12 Options Available Below
Product Details & Documents
Funnels are supplied with a 1:5 PTFE stopcock.
- Heavy wall borosilicate construction
- Ground glass joint
- Teflon® or glass(G) stopcock
- Graduated
Our dropper/addition funnel is specifically designed for the controlled transfer of liquids.
Product Family Options
Product Information
- Capacity[ST] joint sizeStopper diameterAvailabilityPrice
- 60004-258Synthware Pressure Equalizing FunnelSupplier Number: F621410
- 60004-260Synthware Pressure Equalizing FunnelSupplier Number: F621425
- 60004-264Synthware Pressure Equalizing FunnelSupplier Number: F622425
- 60004-266Synthware Pressure Equalizing FunnelSupplier Number: F621460
- 60004-268Synthware Pressure Equalizing FunnelSupplier Number: F621960
- 60004-270Synthware Pressure Equalizing FunnelSupplier Number: F622460
- 60004-272Synthware Pressure Equalizing FunnelSupplier Number: F621125
- 60004-274Synthware Pressure Equalizing FunnelSupplier Number: F629125
- 60004-276Synthware Pressure Equalizing FunnelSupplier Number: F624125
- 60004-278Synthware Pressure Equalizing FunnelSupplier Number: F629250
- 60004-280Synthware Pressure Equalizing FunnelSupplier Number: F624250
- 60004-282Synthware Pressure Equalizing FunnelSupplier Number: F624500