Accessories for Synthware Stopcock, 1:5 PTFE Plug, Straight Bore, Kemtech America
Catalog # 60005-118
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Material:PTFE
- Cat. no.:60005-118
- Stopcock Size:6 mm
- Supplier No.:S181635
Specifications
About this item
Replacement Teflon® straight bore stopcock.
- Teflon® Construction
- Straight bore
- 16/35 Plug size
This replacement kit is complete with one Teflon® plug (1:5 taper), one Teflon® washer, one O-ring.