ContactUsLinkComponent
Synthware Flow Control Adapter, Chromatography, Rodaviss® Joint, Kemtech America
Synthware Flow Control Adapter, Chromatography, Rodaviss® Joint, Kemtech America
Catalog # 60001-748
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Flow Control Adapter, Chromatography, Rodaviss® Joint, Kemtech America
Catalog # 60001-748
Supplier:  Kemtech America
Supplier Number:  C432440
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Environmentally Preferable:
  • Cat. no.:
    60001-748
  • Joint Size:
    24/40
  • Supplier No.:
    C432440
Specifications
About this item
This flow control adapter is designed for use with chromatography columns with a 24/40 or 45/50 inner Rodaviss® joint.
  • Open top screw cap, loosening cap, and Viton® O-ring included
  • With metering plug
Supplied with an open top screw cap, loosening cap, and Viton® O-ring. Adapter has a 0 to 4 mm metering plug.