Black Phenolic Screw Caps, Pulp/Tinfoil Liner, Qorpak®
Supplier: Avantor
Certificates
About this item
Pulp/Tin Foil lined caps offer good chemical resistance and are typically used with solids and powders
These black phenolic screw caps are made of phenolic resin.
Good resistance to hydro-carbons, alcohols, ketones and oils. Not
Frequently Bought Together