ContactUsLinkComponent
Mini Profile™ Capsule Filters, Cytiva (Formerly Pall Lab)
Mini Profile™ Capsule Filters, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
Certificates
undefined
Mini Profile™ Capsule Filters, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
Specifications
  • Catalog No:
  • CA28146-162
  • CA28146-164
  • CA28146-160
  • Effective filtration area:
  • 90 cm2 for Profile Star, 46 cm2 for Profile II
  • 90 cm2 for Profile Star, 46 cm2 for Profile II
  • 90 cm2 for Profile Star, 46 cm2 for Profile II
  • Housing length:
  • 7.7 cm (31/32")
  • 7.7 cm (31/32")
  • 7.7 cm (31/32")
  • Housing Ø:
  • 8.5 cm (311/32")
  • 8.5 cm (311/32")
  • 8.5 cm (311/32")
  • Max. ambient temperature:
  • 50 °C (122 °F) for Profile Star, 30 to 50 °C (86 to 122 °F) for Profile II
  • 50°C (122°F) for Profile Star, 30 to 50°C (86 to 122°F) for Profile II
  • 50 °C (122 °F) for Profile Star, 30 to 50 °C (86 to 122 °F) for Profile II
  • Max. operating pressure:
  • 5.0 bar (75 psi) for Profile Star, 3.4û4.1 bar (50û60 psi) for Profile II
  • 5.0 bar (75 psi) for Profile Star, 3.4–4.1 bar (50–60 psi) for Profile II
  • 5.0 bar (75 psi) for Profile Star, 3.4û4.1 bar (50û60 psi) for Profile II