ContactUsLinkComponent
Whatman™ Autovial Syringeless Filters
Whatman™ Autovial Syringeless Filters
Supplier:  Cytiva
Certificates
undefined
Whatman™ Autovial Syringeless Filters
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • CA28296-030
  • CA28297-004
  • CA28296-024
  • CA28296-012
  • CA11014-020
  • CA28297-124
  • CA28297-120
  • CA28297-122
  • CA28297-118
  • CA28297-106
  • CA28297-108
  • CA80089-522
  • CA21426-503
  • CA28297-104
  • Autoclavable:
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • 121 °C for 20 min
  • Housing material:
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)