Order Entry
Canada
Orders LinkContactUsLinkComponent
 

POLYTRON® PT 1300 D Handheld Homogenizers, Kinematica

Supplier:   Kinematica
POLYTRON® PT 1300 D Handheld Homogenizers, Kinematica
Supplier:  Kinematica
Certificates
POLYTRON® PT 1300 D Handheld Homogenizers, Kinematica
POLYTRON® PT 1300 D Handheld Homogenizers, Kinematica

2 Options Available Below

Product Details & Documents

These handheld homogenizers are used to rapidly break down plant and animal tissue for chemical analysis, prepare emulsions and samples for titrations, and extract pesticides in soil samples

The PT 1300D features digital speed control from 7,000–30,000rpm that maintains set speed (±100rpm) regardless of sample changes. The 100W soft-start motor prevents splashing and sample loss. For operator comfort, the unit also features quiet operation of only 75–85dB at 30,000rpm.

EasyCare™ generators disassemble for cleaning, maintenance, and sterilization, making them ideal for use when cross-contamination is a concern and cleaning is necessary. The generators are also designated "Quick-Connect" as they can be connected/disconnected from the homogenizer quickly and easily. These generators work via the rotor/stator system, in which samples are drawn into the center of the generator tip, vigorously mixed, accelerated, pressed through the narrow gap between the rotor and stator, and forced out between the teeth of the stator. Rotors and stators are cut from a solid block of 316L stainless steel resulting in a stronger, thicker-walled stator tube that extends generator life and prevents deformation.

Homogenizers provide fast, reproducible homogenization, emulsion, dispersion, and mixing, with consistent sample-to-sample uniformity. These units can also be used to homogenize heart, liver, uterus, and smooth muscle for the isolation of mitochondria, intact golgi apparatus, and nuclei, as well as grains and seeds for nutritional analysis.

 : Generators (required for operation) are sold separately. Generators are autoclavable at 121°C (250°F) at 15psi for 15 minutes.

Product Family Options

Homogenizers

Product Information

  • Electrical
    Sample volume
    Availability
    Price
  • 53513-432
    Polytron® PT 1300 D Homogenizer
    Polytron® PT 1300 D Homogenizer
    115 V
    0.05 - 250 ml
    Log in for availability
    Log in for availability
    Each
    Each - $7,215.37
    $7,215.37
    Each
       $7,215.37
     

    Supplier Number: 11010032
  • 53513-434
    Polytron® PT 1300 D Homogenizer
    Polytron® PT 1300 D Homogenizer
    230 V
    0.05 - 250 ml
    Log in for availability
    Log in for availability
    Each
    Each - $7,215.37
    $7,215.37
    Each
       $7,215.37
     

    Supplier Number: 11010030

Generators

Product Information

  • Catalog #
    Description
    For use with
    Diameter ×L
    Sample volume
    Ø
    Availability
    Price
  • PT-DA 03 / 2EC-E050, Saw Tooth
    PT 1200 E, PT 1300 D, PT 2500 E
    3×50 mm
    0.05 - 2 ml
    3 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,487.27
    $3,487.27
    Each
       $3,487.27

Product Information

  • Catalog #
    Description
    For use with
    Diameter ×L
    Sample volume
    Ø
    Availability
    Price
  • PT-DA 12 / 2EC-E157, Saw Tooth
    PT 1200 E, PT 1300 D, PT 2500 E
    12×157 mm
    2 - 250 ml
    12 mm
    Log in for availability
    Log in for availability
    Each
    Each - $4,516.49
    $4,516.49
    Each
       $4,516.49

Product Information

  • Catalog #
    Description
    For use with
    Diameter ×L
    Sample volume
    Ø
    Availability
    Price
  • PT-DA 12 / 2WEC-E157, "W" Style
    PT 1200 E, PT 1300 D, PT 2500 E
    12×157 mm
    2 - 250 ml
    12 mm
    Log in for availability
    Log in for availability
    Each
    Each - $5,025.85
    $5,025.85
    Each
       $5,025.85

Product Information

  • Catalog #
    Description
    For use with
    Diameter ×L
    Sample volume
    Ø
    Availability
    Price
  • PT-DA 07 / 2EC-E107, Saw Tooth
    PT 1200 E, PT 1300 D, PT 2500 E
    7×107 mm
    0.10 - 5 ml
    7 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,857.14
    $3,857.14
    Each
       $3,857.14

Product Information

  • Catalog #
    Description
    For use with
    Diameter ×L
    Sample volume
    Ø
    Availability
    Price
  • PT-DA 05 / 2EC-E085, Saw Tooth
    PT 1200 E, PT 1300 D, PT 2500 E
    5×85 mm
    0.05 - 2 ml
    5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,718.93
    $3,718.93
    Each
       $3,718.93

Product Information

  • Catalog #
    Description
    For use with
    Diameter ×L
    Sample volume
    Ø
    Availability
    Price
  • PT-DA 12 / 2EC-E123, Saw Tooth
    PT 1200 E, PT 1300 D, PT 2500 E
    12×123 mm
    2.00 - 250 ml
    12 mm
    Log in for availability
    Log in for availability
    Each
    Each - $5,078.31
    $5,078.31
    Each
       $5,078.31