GWV High-Capacity Groundwater Sampling Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Certificates
About this item
These disposable filters are designed specifically for the filtration of groundwater prior to dissolved metals analysis.
- Disposable
- For groundwater
- Polypropylene housing
They provide an easy, efficient way to meet filtration requirements of the EPA or other regulatory agency protocols and eliminate the costly decontamination, assembly/disassembly, and multiple filter membrane changes associated with reusable filter holders. Filter media is Versapor® membrane, an acrylic copolymer on a nonwoven support. Polypropylene housing. Inlet/outlet connections: 3.2 mm (¹/₈") MNPT. Effective filtration area: 700 cm²(108¹/₂in.²).
Frequently Bought Together