Carbon Capsule Filter, Cytiva (Formerly Pall Lab)
Catalog #: CA28145-702
Supplier: Cytiva
Specifications
- Description:Carbon Capsule Filter
- Cat. no.:CA28145-702
- Supplier No.:12011
Specifications
About this item
Removes odor, colloids, and organics from liquids using activated carbon.
- Finely divided carbon provides an active surface area of 65000 m²
- Ready to use and disposable
Contains 100 g of acid-rinsed, activated charcoal. Inlet/outlet connections are ³/₈" FNPT. Includes optional hose barb adapters to accept ¹/₂" int.Ø tubing.
Applications:
- For laboratory and reagent-grade water and chemical filtration
- Protect laboratory from organic solvent odor
Customers Who Bought This Also Bought