Whatman™ Papers for Healthcare Applications Grade 470, Whatman products (Cytiva)
Supplier: Cytiva
Certificates
About this item
These filter papers can be used for the absorption of culture media, as a blotting paper, for electrophoresis and amino acid chromatography.
- Soft surface
- For gelatinous samples
Specifications
- Catalog No:
- CA28153-032
- CA28153-030
- Particle retention:
- 20–30 µm
- 20–30 µm
- Porosity:
- Coarse
- Coarse
- Flow rate:
- Fast
- Fast
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together