Order Entry
Canada
ContactUsLinkComponent
Bench Top Biohazard Waste Container
Bench Top Biohazard Waste Container
Catalog # 77677-218
Supplier:  CP Lab Safety
New Product
undefined
Bench Top Biohazard Waste Container
Catalog # 77677-218
Supplier:  CP Lab Safety
Supplier Number:  SWC-002BIO
New Product

Specifications

  • Capacity:
    5 L (1.5 gal)
  • Description:
    Bench top biohazard waste container, with red lid
  • Height:
    11" (if lid closed); 21" (if lid opened)
  • Cat. no.:
    77677-218
  • Liner Material:
    Polyethylene

Specifications

About this item

Bench top biohazard waste container with lid is a convenient way to discard of contaminated biohazardous wastes right where you work. This product is the same as solid waste container but comes with the universal Biohazard label affixed to the container to clearly indicate its contents.

  • This waste container has a small footprint and can easily sit in the corner of a fume hood or bench
  • Made of chemically resistant HDPE and features a detachable base

Features a neon red-orange biohazard label to clearly indicate the nature of its contents. Featuring a detachable base and lid with gasket, our Biohazard waste container is an ideal containment system for contaminated solid wastes, preventing possible cross contamination of products.

The latching lid significantly reduces the emission of biohazardous waste back into the workspace. Ideal if you routinely handle blood borne pathogens, contaminated gloves, wipes, pipets, and other biohazardous disposables.

Comes with a single sample liner bag of 45.72 cm (18") length, 35.56 cm (14") width, 0.05 mm (thick).

Footprint: 13×13" (with base), 7×7" (without base).