Hepatitis C virus (HCV) Recombinant NS5
Catalog # 77682-570
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/peptide type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein synonyms:non structural protein 5|nonstructural antigen 5|nonstructural protein 5
- Protein/peptide name:NS5
- Purity:HCV-NS5 HRP protein is >95% pure as determined by 10% page (coomassie staining)
- Endotoxin level:Very low
- Formulation:50 mM tris pH-8 and amp 5 mM EDTA
- Shipping temperature:Dry Ice
- Cat. no.:77682-570
Specifications
About this item
Recombinant HCV NS5 Antigen, Biotin-Labled.