Hepatitis C virus (HCV) Recombinant Non-structural protein 5A
Catalog # 77682-538
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/peptide type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein synonyms:p56|NS5a immunodominant regions|NS5A
- Protein/peptide name:Non-structural protein 5A
- Purity:HCV-NS5 protein is >95% pure as determined by 10% page (coomassie staining)
- Endotoxin level:Very low
- Formulation:1.5M Urea, 25 mM tris-HCl pH 8, 50% glycerol and 0.2% Triton-X
- Shipping temperature:Dry Ice
- Cat. no.:77682-538
Specifications
About this item
Recombinant HCV NS5A Antigen, GST-tagged.