Hepatitis C virus (HCV) Recombinant NS4
77682-528
New Product
:
- Pack type:Vial
- Protein/peptide type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein synonyms:nonstructural antigen 4|p8|Non structural protein 4A|p27|NS4B|Non structural protein 4B|NS4 Mosaic|NS4A
- Protein/peptide name:NS4
- Purity:Protein is >95% pure as determined by 10% page (coomassie staining)
- Endotoxin level:Very low
- Formulation:1.5 M urea, 25 mM tris-HCl, pH 8.0, 0.2% Triton-X and amp 50% glycerol
- Shipping temperature:Dry Ice
- Cat. no.:77682-528
Recombinant HCV Genotype-1 NS4 Mosaic Antigen, GST-tagged.