Hepatitis C virus (HCV) Recombinant NS4
77682-526
New Product
:
- Pack type:Vial
- Protein/peptide type:Recombinant
- Species:Hepatitis C virus (HCV)
- Protein synonyms:nonstructural antigen 4|p8|Non structural protein 4A|p27|NS4B|Non structural protein 4B|NS4 Mosaic|NS4A
- Protein/peptide name:NS4
- Purity:Protein is >95% pure as determined by 10% page (coomassie staining)
- Molecular weight:66 kDa
- Endotoxin level:Very low
- Formulation:8 M Urea, 20 mM tris, 10 mM DTT, 1 mM EDTA and 0.5 Tween-20, pH-9
- Shipping temperature:Dry Ice
- Cat. no.:77682-526
Recombinant HCV NS4 Mosaic Antigen, His-tagged.