N. meningitidis Recombinant Neisseria meningitidis Serogroup B strain NZ98/254 outer membrane vesicle
Catalog # 77682-462
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Protein/peptide type:Recombinant
- Species:N. meningitidis
- Protein/peptide name:Neisseria meningitidis Serogroup B strain NZ98/254 outer membrane vesicle
- Endotoxin level:Very low
- Shipping temperature:Dry Ice
- Cat. no.:77682-462
Specifications
About this item
Recombinant Neisseria meningitidis Serogroup B strain NZ98/254 outer membrane vesicle.