Rabies virus Rabies virus strain flury lep antigen (propiolactone inactivated)
Catalog # 77682-178
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Species:Rabies virus
- Protein synonyms:Rabies virus vaccine flury-lep strain|Rabies virus strain flury lep antigen (propiolactone inactivated)
- Protein/peptide name:Rabies virus strain flury lep antigen (propiolactone inactivated)
- Endotoxin level:very low
- Grade:Ultrapure grade
- Shipping temperature:Dry Ice
- Cat. no.:77682-178
Specifications
About this item
Rabies virus inactivated antigen, A.