Horse Recombinant RE VEGF-165 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
New Product
Certificates
About this item
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant. The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
- High quality
- Low endotoxin
- Affordable
VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant. The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Horse
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: HUVEC Proliferation. ED50 ≤10 ng/ml. bioactivity ≥1.0×10⁵ units/mg
- Gene ID:Q9GKR0
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:glioma-derived endothelial cell mitogen|VEGF-A|VPF
- Accession no.:Q9GKR0
- Protein/peptide name:RE VEGF-165
- Purity:≥95%
- Molecular weight:19.3/38.6 kDa (165/330 aa), dimer
- Sequence:MAPMAEGEHK THEVVKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCGGC
CNDEGLECVP TAEFNITMQI MRIKPHQSQH IGEMSFLQHS KCECRPKKDK ARQENPCGPC
SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR - Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together