Anti-CDC7 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, IHC:1:25-1:100
Type:
Antigen: CDC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77706-664
- 77706-663
- Antigen symbol:
- CDC7
- CDC7
- Antigen synonyms:
- Cdc7 like 1|CDC7-related kinase|CDC7 cell division cycle 7 like 1|MGC117361|CDC7L 1|MGC126238|Cell division cycle 7 kinase|MGC126237|CDC7L1|Cell division cycle 7 like protein 1|HuCDC 7|Cdc7 Kinase|CDC7|CDC7 cell division cycle 7|Cell division cycle 7 related protein kinase|Cdc 7|CDC7 L1|CDC7_HUMAN|HsCDC 7|Cell division cycle 7 homolog|HsCdc7|Hsk1|Hsk 1|Cell division cycle 7-related protein kinase|huCdc7|CDC7 related kinase
- Cdc7 like 1|CDC7-related kinase|CDC7 cell division cycle 7 like 1|MGC117361|CDC7L 1|MGC126238|Cell division cycle 7 kinase|MGC126237|CDC7L1|Cell division cycle 7 like protein 1|HuCDC 7|Cdc7 Kinase|CDC7|CDC7 cell division cycle 7|Cell division cycle 7 related protein kinase|Cdc 7|CDC7 L1|CDC7_HUMAN|HsCDC 7|Cell division cycle 7 homolog|HsCdc7|Hsk1|Hsk 1|Cell division cycle 7-related protein kinase|huCdc7|CDC7 related kinase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00311
- O00311
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CDC7
- Synthetic peptide of Human CDC7
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together