Anti-PPBP Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPBP
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77706-048
- 77706-049
- Antigen symbol:
- PPBP
- PPBP
- Antigen synonyms:
- CTAP 3|Beta-TG|CTAP III|CTAPIII|Connective tissue activating peptide III|beta-1|SCYB7|Chemokine (C X C motif) ligand 7|CTAP3|CXCL 7|TC2|Platelet basic protein|TGB|NAP-2(1-63)|Macrophage-derived growth factor|Beta TG |Thrombocidin 1|CTAP-III|MDGF|B TG1|LDGF|PBP|NAP-2(1-66)|PPBP|Thromboglobulin|Neutrophil activating peptide 2|Leukocyte derived growth factor|THBGB1|LA PF 4|CXCL7_HUMAN|Leukocyte-derived growth factor|Small inducible cytokine subfamily B member 7|Thrombocidin 2|Low-affinity platelet factor IV|LA-PF4|Neutrophil-activating peptide 2(1-63)|NAP-2(74)|Pro platelet basic protein|Beta thromboglobulin|THBGB|TC1|TGB1|C-X-C motif chemokine 7|Small-inducible cytokine B7|NAP-2|NAP 2|CXC chemokine ligand 7|CXCL7|CTAP-III(1-81)|Pro platelet basic protein (chemokine (C-X-C motif) ligand 7)|NAP-2(73)
- CTAP 3|Beta-TG|CTAP III|CTAPIII|Connective tissue activating peptide III|beta-1|SCYB7|Chemokine (C X C motif) ligand 7|CTAP3|CXCL 7|TC2|Platelet basic protein|TGB|NAP-2(1-63)|Macrophage-derived growth factor|Beta TG |Thrombocidin 1|CTAP-III|MDGF|B TG1|LDGF|PBP|NAP-2(1-66)|PPBP|Thromboglobulin|Neutrophil activating peptide 2|Leukocyte derived growth factor|THBGB1|LA PF 4|CXCL7_HUMAN|Leukocyte-derived growth factor|Small inducible cytokine subfamily B member 7|Thrombocidin 2|Low-affinity platelet factor IV|LA-PF4|Neutrophil-activating peptide 2(1-63)|NAP-2(74)|Pro platelet basic protein|Beta thromboglobulin|THBGB|TC1|TGB1|C-X-C motif chemokine 7|Small-inducible cytokine B7|NAP-2|NAP 2|CXC chemokine ligand 7|CXCL7|CTAP-III(1-81)|Pro platelet basic protein (chemokine (C-X-C motif) ligand 7)|NAP-2(73)
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02775
- P02775
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Platelet basic protein (59-125AA)
- Recombinant Human Platelet basic protein (59-125AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together