Anti-CCL16 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CCL16
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77706-018
- 77706-019
- Antigen symbol:
- CCL16
- CCL16
- Antigen synonyms:
- NCC-4|CCL16|SCYL4|Small inducible cytokine subfamily A (Cys Cys) member 16|Chemokine LEC|Liver expressed chemokine|ILINCK|CKb12|Lymphocyte and monocyte chemoattractant|LCC1|Small-inducible cytokine A16|Monotactin 1|Chemokine (C-C motif) ligand 16|Monotactin-1|HCC4|IL10 inducible chemokine|Liver-expressed chemokine|MTN-1|CCL16_HUMAN|C-C motif chemokine 16|Mtn1|New CC chemokine 4|SCYA16|Liver CC chemokine 1 precursor|NCC4|Small inducible cytokine A16 precursor|HCC-4|Chemokine CC-4|LCC-1|LMC
- NCC-4|CCL16|SCYL4|Small inducible cytokine subfamily A (Cys Cys) member 16|Chemokine LEC|Liver expressed chemokine|ILINCK|CKb12|Lymphocyte and monocyte chemoattractant|LCC1|Small-inducible cytokine A16|Monotactin 1|Chemokine (C-C motif) ligand 16|Monotactin-1|HCC4|IL10 inducible chemokine|Liver-expressed chemokine|MTN-1|CCL16_HUMAN|C-C motif chemokine 16|Mtn1|New CC chemokine 4|SCYA16|Liver CC chemokine 1 precursor|NCC4|Small inducible cytokine A16 precursor|HCC-4|Chemokine CC-4|LCC-1|LMC
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15467
- O15467
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human C-C motif chemokine 16 protein (24-120AA)
- Recombinant Human C-C motif chemokine 16 protein (24-120AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together