Anti-PLA2G7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLA2G7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77705-965
- 77689-095
- Antigen symbol:
- PLA2G7
- PLA2G7
- Antigen synonyms:
- 1 alkyl 2 acetylglycerophosphocholine esterase|PAF 2-acylhydrolase|PAFAD|PLA2G7|2-acetyl-1-alkylglycerophosphocholine esterase|LDL PLA2|Platelet activating factor acetylhydrolase 2|gVIIA PLA2|LDL-associated phospholipase A2|PAFAH|Group VIIA phospholipase A2|Platelet activating factor acetylhydrolase|PAFA_HUMAN|PAF 2 acylhydrolase LDL associated phospholipase A2|gVIIA-PLA2|LDL PLA(2)|2 acetyl 1 alkylglycerophosphocholine esterase|Lipoprotein associated phospholipase A2|Phospholipase A2, group VII (platelet activating factor acetylhydrolase, plasma)|LP PLA2|1-alkyl-2-acetylglycerophosphocholine esterase|Platelet-activating factor acetylhydrolase precursor|LDL associated phospholipase A2|PAF acetylhydrolase|EC 3.1.1.47 PAF acetylhydrolase PAF 2-acylhydrolase LDL associated phospholipase A2|Group-VIIA phospholipase A2|Platelet-activating factor acetylhydrolase|PAF 2 acylhydrolase|LDL-PLA(2)
- 1 alkyl 2 acetylglycerophosphocholine esterase|PAF 2-acylhydrolase|PAFAD|PLA2G7|2-acetyl-1-alkylglycerophosphocholine esterase|LDL PLA2|Platelet activating factor acetylhydrolase 2|gVIIA PLA2|LDL-associated phospholipase A2|PAFAH|Group VIIA phospholipase A2|Platelet activating factor acetylhydrolase|PAFA_HUMAN|PAF 2 acylhydrolase LDL associated phospholipase A2|gVIIA-PLA2|LDL PLA(2)|2 acetyl 1 alkylglycerophosphocholine esterase|Lipoprotein associated phospholipase A2|Phospholipase A2, group VII (platelet activating factor acetylhydrolase, plasma)|LP PLA2|1-alkyl-2-acetylglycerophosphocholine esterase|Platelet-activating factor acetylhydrolase precursor|LDL associated phospholipase A2|PAF acetylhydrolase|EC 3.1.1.47 PAF acetylhydrolase PAF 2-acylhydrolase LDL associated phospholipase A2|Group-VIIA phospholipase A2|Platelet-activating factor acetylhydrolase|PAF 2 acylhydrolase|LDL-PLA(2)
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13093
- Q13093
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Platelet-activating factor acetylhydrolase protein (49-441AA)
- Recombinant Human Platelet-activating factor acetylhydrolase protein (49-441AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together