Anti-CYP2A7 Rabbit Polyclonal Antibody
Catalog # 77702-000
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CYP2A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:P20853
- Antigen synonyms:CYP2A7Cytochrome P450 2A7|EC 1.14.14.1|CYPIIA7|Cytochrome P450 IIA4
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CYP2A6/7.
- Purification:Antigen affinity purified
- Cat. no.:77702-000
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: CYP2A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human