Anti-PNCK Rabbit Polyclonal Antibody
Catalog # 77701-893
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:PNCK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q6P2M8
- Antigen synonyms:CaM kinase IB|CaMKI-beta|CaM kinase I beta|CaM-KI beta|Calcium/calmodulin dependent protein kinase type 1B|Calcium/calmodulin-dependent protein kinase type 1B|CaMKI beta|PNCK|Pregnancy up-regulated non-ubiquitously-expressed CaM kinase|EC 2.7.11.17|CaMK1b|MGC45419|Pregnancy up regulated non ubiquitously expressed CaM kinase|CaM KI beta|KCC1B_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CaMKIβ.
- Purification:Antigen affinity purified
- Cat. no.:77701-893
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: PNCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat